3-Amino-5-chloro-2-hydroxyphenylbenzenecarboxylate 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-5-chloro-2-hydroxyphenyl benzoate

OR40057-1G OR40057-500MG
APOSOR40057-1GEA 677 EUR
APOSOR40057-1G APOSOR40057-500MG
3-Amino-5-chloro-2-hydroxyphenylbenzenecarboxylate 95+%
3-Amino-5-chloro-2-hydroxyphenylbenzenecarboxylate
Formula: C₁₃H₁₀ClNO₃
MW: 263,68 g/mol
Melting Pt: 169…170 °C
Storage Temperature: Ambient
MDL Number: MFCD16140259
CAS Number: 1221791-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR