Methyl-4-iodo-3-methoxybenzoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 4-iodo-3-methoxybenzoate

OR400591-1G OR400591-5G OR400591-250MG
APOSOR400591-1GEA 46.4 EUR
APOSOR400591-1G APOSOR400591-5G APOSOR400591-250MG
Methyl-4-iodo-3-methoxybenzoate 98%
Methyl 4-iodo-3-methoxybenzoate
Formula: C₉H₉IO₃
MW: 292.0704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203960
CAS Number: 35387-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR