4-Bromo-1-(bromomethyl)-2-nitrobenzene
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-Bromo-2-nitrobenzyl bromide
OR400618-1G
OR400618-25G
OR400618-5G
APOSOR400618-1GEA
66.7
EUR
APOSOR400618-1G
APOSOR400618-25G
APOSOR400618-5G
4-Bromo-1-(bromomethyl)-2-nitrobenzene
4-Bromo-1-(bromomethyl)-2-nitrobenzene
Formula:
C₇H₅Br₂NO₂ Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD07779494 CAS Number: 82420-34-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...