1-Bromo-3-chloro-5-iodobenzene
Supplier: Apollo Scientific
OR400623-25G
APOSOR400623-25GEA
119
EUR
APOSOR400623-25G
1-Bromo-3-chloro-5-iodobenzene
1-Bromo-3-chloro-5-iodobenzene
Formula:
C₆H₃BrClI MW: 317,3500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07778998 CAS Number: 13101-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...