Ethyl-4-chloronicotinate
Supplier: Apollo Scientific
|
Warning
|
OR40065-1G
OR40065-250MG
OR40065-5G
APOSOR40065-1GEA
568
EUR
APOSOR40065-1G
APOSOR40065-250MG
APOSOR40065-5G
Ethyl-4-chloronicotinate
Ethyl-4-chloronicotinate
Formula:
C₈H₈ClNO₂ MW: 185,61 g/mol Boiling Pt: 85 °C/1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD05664028 CAS Number: 37831-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...