3-Chlorosalicylic acid 97%

Supplier: Apollo Scientific
Warning

OR400661-1G OR400661-25G OR400661-5G
APOSOR400661-1GEA 21.9 EUR
APOSOR400661-1G APOSOR400661-25G APOSOR400661-5G
3-Chlorosalicylic acid 97%
3-Chlorosalicylic acid
Formula: C₇H₅ClO₃
MW: 172,57 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00010390
CAS Number: 1829-32-9
EINECS: 217-379-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR