3-Chloro-4-iodophenol

Supplier: Apollo Scientific

OR400917-1G OR400917-250MG
APOSOR400917-1GEA 309 EUR
APOSOR400917-1G APOSOR400917-250MG
3-Chloro-4-iodophenol
3-Chloro-4-iodophenol
Formula: C₆H₄ClIO
Storage Temperature: Ambient
MDL Number: MFCD17000018
CAS Number: 855403-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR