2-Chloro-3-nitrobenzaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
OR401003-1G
OR401003-5G
APOSOR401003-1GEA
125
EUR
APOSOR401003-1G
APOSOR401003-5G
2-Chloro-3-nitrobenzaldehyde 95%
2-Chloro-3-nitrobenzaldehyde
Formula:
C₇H₄ClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD08236804 CAS Number: 58755-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...