2-Iodophenyl isocyanate

Supplier: Apollo Scientific
Danger

OR401067-1G OR401067-25G OR401067-5G
APOSOR401067-1GEA 58 EUR
APOSOR401067-1G APOSOR401067-25G APOSOR401067-5G
2-Iodophenyl isocyanate
2-Iodophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄INO
MW: 245.02 g/mol
Boiling Pt: 245 °C
Density: 1.891 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00673044
CAS Number: 128255-31-2
UN: 2206
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR