3-Iodophenyl isocyanate

Supplier: Apollo Scientific
Danger

OR401068-1G OR401068-25G OR401068-5G
APOSOR401068-1GEA 72.5 EUR
APOSOR401068-1G APOSOR401068-25G APOSOR401068-5G
3-Iodophenyl isocyanate
3-Iodophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄INO
MW: 245.02 g/mol
Boiling Pt: 55 °C/0.23mmHg
Density: 1.86 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037088
CAS Number: 23138-56-9
UN: 2206
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR