4-Methyl-2-nitroanisole 98+%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Methoxy-3-nitrotoluene, 4-Methoxy-4-nitrotoluene, Methyl 2-nitro-p-tolyl ether, 1-Methoxy-4-methyl-2-nitrobenzene, 4-Methyl-2-nitroanisole

OR40113-100G OR40113-25G
APOSOR40113-100GEA 116 EUR
APOSOR40113-100G APOSOR40113-25G
4-Methyl-2-nitroanisole 98+%
4-Methyl-2-nitroanisole
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 154 °C/14mmHg
Melting Pt: 44447 °C
Density: 1.205 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00024540
CAS Number: 119-10-8
EINECS: 204-296-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR