4-Bromo-5-methyl-1H-indazole

Supplier: Apollo Scientific
Warning

OR40118-5G
APOSOR40118-5GEA 94.4 EUR
APOSOR40118-5G
4-Bromo-5-methyl-1H-indazole
4-Bromo-5-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211,0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044156
CAS Number: 926922-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR