3-(4-Morpholinylcarbonyl)phenylboronic acid 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3-(morpholine-4-carbonyl)phenyl)boronic acid, 3-(Morpholine-4-carbonyl)phenylboronic acid, 3-(4-Morpholinylcarbonyl)benzeneboronic acid
OR4012-1G
OR4012-5G
APOSOR4012-1GEA
173
EUR
APOSOR4012-1G
APOSOR4012-5G
3-(4-Morpholinylcarbonyl)phenylboronic acid 96%
3-(4-Morpholinylcarbonyl)phenylboronic acid
Formula:
C₁₁H₁₄BNO₄ MW: 235,05 g/mol Melting Pt: 124…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115687 CAS Number: 723281-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...