3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid 96%

Supplier: Apollo Scientific
Warning

Synonyms: (3-((2-(Dimethylamino)ethyl)carbamoyl)phenyl)boronic acid

OR4014-1G
APOSOR4014-1GEA 317 EUR
APOSOR4014-1G
3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid 96%
3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid
Formula: C₁₁H₁₇BN₂O₃
MW: 236,08 g/mol
Melting Pt: 42…45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115706
CAS Number: 850567-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR