5-Bromobenzotriazole 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-1H-1,2,3-benzotriazole, 5-Bromo-1H-benzotriazole
OR40184-1G
OR40184-5G
APOSOR40184-1GEA
122
EUR
APOSOR40184-1G
APOSOR40184-5G
5-Bromobenzotriazole 95%
5-Bromobenzotriazole
Formula:
C₆H₄BrN₃ MW: 198,02 g/mol Melting Pt: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00464510 CAS Number: 32046-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...