Chlorotriethylsilane 99%
Supplier: Apollo Scientific
|
Danger
|
OR40296-100ML
OR40296-500ML
APOSOR40296-100MLEA
256
EUR
APOSOR40296-100ML
APOSOR40296-500ML
Chlorotriethylsilane 99%
Chlorotriethylsilane
Formula:
C₆H₁₅ClSi Boiling Pt: 142…147 °C Melting Pt: –75 °C Density: 0,898 g/cm³ (25 °C) Flash Pt: 30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000507 CAS Number: 994-30-9 EINECS: 213-615-6 UN: 2986 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...