1-Bromo-3-iodo-5-nitrobenzene 95%
Supplier: Apollo Scientific
|
Warning
|
OR40409-1G
OR40409-250MG
APOSOR40409-1GEA
512
EUR
APOSOR40409-1G
APOSOR40409-250MG
1-Bromo-3-iodo-5-nitrobenzene 95%
1-Bromo-3-iodo-5-nitrobenzene
Formula:
C₆H₃BrINO₂ MW: 327,9020 g/mol Melting Pt: 97…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD13194576 CAS Number: 861601-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...