Triphenylsulphonium chloride 95%

Supplier: Apollo Scientific
Warning

OR40449-1G OR40449-5G
APOSOR40449-1GEA 21.9 EUR
APOSOR40449-1G APOSOR40449-5G
Triphenylsulphonium chloride 95%
Triphenylsulphonium chloride
Formula: (C₆H₅)₃SCl
MW: 298,84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031716
CAS Number: 4270-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR