4,6-Dimethoxypyrimidine-2-carboxylic acid 95+%

Supplier: Apollo Scientific
Warning

OR4045-1G OR4045-5G
APOSOR4045-1GEA 94.4 EUR
APOSOR4045-1G APOSOR4045-5G
4,6-Dimethoxypyrimidine-2-carboxylic acid 95+%
4,6-Dimethoxypyrimidine-2-carboxylic acid
Formula: C₇H₈N₂O₄
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD03645365
CAS Number: 128276-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR