6-Methoxyquinaldine

Supplier: Apollo Scientific
Warning

OR40463-1G OR40463-5G
APOSOR40463-1GEA 21.9 EUR
APOSOR40463-1G APOSOR40463-5G
6-Methoxyquinaldine
6-Methoxyquinaldine
Formula: C₁₁H₁₁NO
Melting Pt: 62…66 °C
Density: 0,973 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006761
CAS Number: 1078-28-0
EINECS: 214-080-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR