2-Amino-3-formylpyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-3-pyridinecarboxaldehyde, 2-Aminonicotinaldehyde, 2-Aminopyridine-3-carboxaldehyde

OR40471-25G OR40471-5G OR40471-1G
APOSOR40471-25GEA 82.9 EUR
APOSOR40471-25G APOSOR40471-5G APOSOR40471-1G
2-Amino-3-formylpyridine 98%
2-Amino-3-formylpyridine
Formula: C₆H₆N₂O
MW: 122,13 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01830382
CAS Number: 7521-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR