4-Amino-3-formylpyridine 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Aminopyridine-3-carboxaldehyde, 4-Aminonicotinaldehyde
OR4048-1G
OR4048-250MG
APOSOR4048-1GEA
16.1
EUR
APOSOR4048-1G
APOSOR4048-250MG
4-Amino-3-formylpyridine 99%
4-Amino-3-formylpyridine
Formula:
C₆H₆N₂O MW: 122,13 g/mol Melting Pt: 108…113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04038972 CAS Number: 42373-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...