Tetrachloro-o-benzoquinone

Supplier: Apollo Scientific
Warning

Synonyms: 3,4,5,6-Tetrachloro-1,2-benzoquinone, Tetrachloro-1,2-benzoquinone, 2-Chloranil, Tetrachloro-o-benzoquinone, o-Chloranil, 3,4,5,6-Tetrachlorocyclohexa-3,5-diene-1,2-dione

OR40496-25G OR40496-5G
APOSOR40496-25GEA 452 EUR
APOSOR40496-25G APOSOR40496-5G
Tetrachloro-o-benzoquinone
Tetrachloro-o-benzoquinone
Formula: C₆Cl₄O₂
MW: 245,88 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00001646
CAS Number: 2435-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR