5-Bromo-1H-inden-2(3H)-one
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromoindan-2-one
OR40513-100MG
OR40513-250MG
OR40513-1G
APOSOR40513-100MGEA
81.2
EUR
APOSOR40513-100MG
APOSOR40513-250MG
APOSOR40513-1G
5-Bromo-1H-inden-2(3H)-one
5-Bromo-1H-inden-2(3H)-one
Formula:
C₉H₇BrO MW: 211,0600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07778391 CAS Number: 174349-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...