7-Bromo-1-benzothiophene-2-carboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 7-Bromobenzo[b]thiophene-2-carboxylic acid

OR40543-5G
APOSOR40543-5GEA 98.6 EUR
APOSOR40543-5G
7-Bromo-1-benzothiophene-2-carboxylic acid 98%
7-Bromo-1-benzothiophene-2-carboxylic acid
Formula: C₉H₅BrO₂S
MW: 257,11 g/mol
Melting Pt: >275 °C
Storage Temperature: Ambient
MDL Number: MFCD12032270
CAS Number: 19075-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR