4-Amino-2-methylbenzoic acid

Supplier: Apollo Scientific

Synonyms: 4-Amino-o-toluic acid

OR40554-100G OR40554-25G OR40554-5G
APOSOR40554-100GEA 56.8 EUR
APOSOR40554-100G APOSOR40554-25G APOSOR40554-5G
4-Amino-2-methylbenzoic acid
4-Amino-2-methylbenzoic acid
Formula: C₈H₉NO₂
MW: 151,16 g/mol
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD06656133
CAS Number: 2486-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR