2,3,5-Trimethylphenol 98%

Supplier: Apollo Scientific
Danger

OR40560-500G
APOSOR40560-500GEA 169 EUR
APOSOR40560-500G
2,3,5-Trimethylphenol 98%
2,3,5-Trimethylphenol
Formula: C₉H₁₂O
MW: 136,19 g/mol
Boiling Pt: 230…231 °C
Melting Pt: 92…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002228
CAS Number: 697-82-5
EINECS: 211-806-9
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR