1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt 98+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,3,6,8-Pyrenetetrasulphonic acid tetrasodium salt
OR40562-25G
OR40562-100G
OR40562-500G
APOSOR40562-25GEA
21.9
EUR
APOSOR40562-25G
APOSOR40562-100G
APOSOR40562-500G
1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt 98+%
1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt
Formula:
C₁₆H₆Na₄O₁₂S₄ Storage Temperature: Ambient |
MDL Number:
MFCD09263184 CAS Number: 59572-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...