3-Chloropicolinic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Chloropyridine-2-carboxylic acid
OR40652-1G
OR40652-25G
OR40652-5G
APOSOR40652-1GEA
51
EUR
APOSOR40652-1G
APOSOR40652-25G
APOSOR40652-5G
3-Chloropicolinic acid
3-Chloropicolinic acid
Formula:
C₆H₄ClNO₂ MW: 157,56 g/mol Melting Pt: 131 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01646325 CAS Number: 57266-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...