2-Bromo-5-iodophenol 97%

Supplier: Apollo Scientific
Warning

OR40667-1G OR40667-250MG OR40667-5G
APOSOR40667-1GEA 24.8 EUR
APOSOR40667-1G APOSOR40667-250MG APOSOR40667-5G
2-Bromo-5-iodophenol 97%
2-Bromo-5-iodophenol
Formula: C₆H₄BrIO
MW: 298,9000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907878
CAS Number: 932372-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR