2-Bromo-4-iodobenzaldehyde 97%

Supplier: Apollo Scientific
Warning

OR40673-1G OR40673-5G
APOSOR40673-1GEA 74.2 EUR
APOSOR40673-1G APOSOR40673-5G
2-Bromo-4-iodobenzaldehyde 97%
2-Bromo-4-iodobenzaldehyde
Formula: C₇H₄BrIO
MW: 310,9200 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD07779025
CAS Number: 261903-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR