α,4-Dichloro-α-phenyltoluene
Supplier: Apollo Scientific
|
Danger
|
OR40704-100G
OR40704-1KG
OR40704-500G
APOSOR40704-100GEA
58
EUR
APOSOR40704-100G
APOSOR40704-1KG
APOSOR40704-500G
α,4-Dichloro-α-phenyltoluene
α,4-Dichloro-α-phenyltoluene
Formula:
C₁₃H₁₀Cl₂ Boiling Pt: 159…160 °C/2mmHg Density: 1,239 g/cm³ (25 °C) Flash Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000856 CAS Number: 134-83-8 EINECS: 205-158-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...