(m-Acrylamidophenyl)boronic acid 95%
Supplier: Apollo Scientific
|
Warning
|
OR40723-1G
OR40723-5G
APOSOR40723-1GEA
39.4
EUR
APOSOR40723-1G
APOSOR40723-5G
(m-Acrylamidophenyl)boronic acid 95%
(m-Acrylamidophenyl)boronic acid
Formula:
C₉H₁₀BNO₃ Melting Pt: 148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09025755 CAS Number: 99349-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...