Isothiazole-3-carboxylic acid 97%

Supplier: Apollo Scientific
Warning

OR40803-250MG OR40803-100MG OR40803-1G
APOSOR40803-250MGEA 261 EUR
APOSOR40803-250MG APOSOR40803-100MG APOSOR40803-1G
Isothiazole-3-carboxylic acid 97%
Isothiazole-3-carboxylic acid
Formula: C₄H₃NO₂S
Melting Pt: 135…137 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834765
CAS Number: 4576-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR