4-Bromo-1-methylpyridin-2(1H)-one 98%

Supplier: Apollo Scientific
Warning

OR41149-1G OR41149-5G OR41149-25G
APOSOR41149-1GEA 75.7 EUR
APOSOR41149-1G APOSOR41149-5G APOSOR41149-25G
4-Bromo-1-methylpyridin-2(1H)-one 98%
4-Bromo-1-methylpyridin-2(1H)-one
Formula: C₆H₆BrNO
MW: 188,0200 g/mol
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD11226870
CAS Number: 214342-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR