5-Aminopyridin-3-ol 97+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Amino-5-hydroxypyridine
OR41178-1G
OR41178-5G
APOSOR41178-1GEA
181
EUR
APOSOR41178-1G
APOSOR41178-5G
5-Aminopyridin-3-ol 97+%
5-Aminopyridin-3-ol
Formula:
C₅H₆N₂O MW: 110,1139 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04114213 CAS Number: 3543-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...