Uracil-5-carboxaldehyde 98%
Supplier: Apollo Scientific
Synonyms:
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, 5-Formyluracil
OR4144-1G
OR4144-250MG
APOSOR4144-1GEA
0
EUR
APOSOR4144-1G
APOSOR4144-250MG
Uracil-5-carboxaldehyde 98%
Uracil-5-carboxaldehyde
Formula:
C₅H₄N₂O₃ MW: 140,10 g/mol Melting Pt: >300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00192185 CAS Number: 1195-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...