1-Hexylpiperazine

Supplier: Apollo Scientific
Warning

OR4146-1G OR4146-250MG
APOSOR4146-1GEA 24.8 EUR
APOSOR4146-1G APOSOR4146-250MG
1-Hexylpiperazine
1-Hexylpiperazine
Formula: C₁₀H₂₂N₂
Boiling Pt: 98…103 °C/10mmHg
Density: 0,877 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01075185
CAS Number: 51619-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR