1-Pentylpiperazine

Supplier: Apollo Scientific
Warning

OR4168-1G OR4168-250MG
APOSOR4168-1GEA 45.1 EUR
APOSOR4168-1G APOSOR4168-250MG
1-Pentylpiperazine
1-Pentylpiperazine
Formula: C₉H₂₀N₂
Boiling Pt: 88…91 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD02093555
CAS Number: 50866-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR