4-Phenylpiperidine 97%
Supplier: Apollo Scientific
|
Warning
|
OR4179-10G
OR4179-5G
OR4179-1G
APOSOR4179-10GEA
66.7
EUR
APOSOR4179-10G
APOSOR4179-5G
APOSOR4179-1G
4-Phenylpiperidine 97%
4-Phenylpiperidine
Formula:
C₁₁H₁₅N MW: 161.25 g/mol Boiling Pt: 255…257 °C/727mmHg Melting Pt: 61…65 °C Density: 0.967 Storage Temperature: Ambient |
MDL Number:
MFCD00006002 CAS Number: 771-99-3 EINECS: 212-243-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...