4-Methoxystyrene

Supplier: Apollo Scientific
Warning

OR42010-1G OR42010-5G
APOSOR42010-1GEA 29 EUR
APOSOR42010-1G APOSOR42010-5G
4-Methoxystyrene
4-Methoxystyrene
Formula: C₉H₁₀O
Boiling Pt: 41…42 °C/0,5mmHg
Density: 1,002 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008619
CAS Number: 637-69-4
EINECS: 211-298-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR