1,7-Octadiyne 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Octa-1,7-diyne
OR42022-10G
OR42022-50G
APOSOR42022-10GEA
125
EUR
APOSOR42022-10G
APOSOR42022-50G
1,7-Octadiyne 98%
1,7-Octadiyne
Formula:
C₈H₁₀ MW: 106,17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00008580 CAS Number: 871-84-1 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...