4-Chloro-2-iodo-6-methylaniline 95+%

Supplier: Apollo Scientific
Danger

OR42163-1G OR42163-500MG OR42163-5G
APOSOR42163-1GEA 292 EUR
APOSOR42163-1G APOSOR42163-500MG APOSOR42163-5G
4-Chloro-2-iodo-6-methylaniline 95+%
4-Chloro-2-iodo-6-methylaniline
Formula: C₇H₇CllN
Melting Pt: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD16622764
CAS Number: 69705-29-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR