8-Chloroquinoline 98%
Supplier: Apollo Scientific
|
Warning
|
OR42189-100G
APOSOR42189-100GEA
165
EUR
APOSOR42189-100G
8-Chloroquinoline 98%
8-Chloroquinoline
Formula:
C₉H₆ClN Boiling Pt: 288 °C Melting Pt: –20 °C Density: 1,28 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047618 CAS Number: 611-33-6 EINECS: 210-265-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...