1-Bromo-2-chloro-5-nitrobenzene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-4-chloronitrobenzene

OR4221-25G OR4221-100G
APOSOR4221-25GEA 65.4 EUR
APOSOR4221-25G APOSOR4221-100G
1-Bromo-2-chloro-5-nitrobenzene 98%
1-Bromo-2-chloro-5-nitrobenzene
Formula: C₆H₃BrClNO₂
MW: 236,4500 g/mol
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00100437
CAS Number: 16588-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR