Bis(2-bromoethyl)ether 95%

Supplier: Apollo Scientific
Danger

Synonyms: 1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether

OR42237-25G OR42237-5G
APOSOR42237-25GEA 252 EUR
APOSOR42237-25G APOSOR42237-5G
Bis(2-bromoethyl)ether 95%
Bis(2-bromoethyl) ether
Formula: C₄H₈Br₂O
MW: 231,93 g/mol
Boiling Pt: 92…93 °C/12mmHg
Density: 1,845 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00039196
CAS Number: 5414-19-7
EINECS: 226-504-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR