6-Aminopyridin-2-ol, Technical Grade
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
6-Hydroxypyridin-2-amine, 6-Aminopyridin-2-ol
OR4269-5G
OR4269-1G
OR4269-25G
APOSOR4269-5GEA
48
EUR
APOSOR4269-5G
APOSOR4269-1G
APOSOR4269-25G
6-Aminopyridin-2-ol, Technical Grade
6-Aminopyridin-2-ol
Formula:
C₅H₆N₂O MW: 110,11 g/mol Melting Pt: 210…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039717 CAS Number: 5154-00-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...