1-Bromo-3-nitrobenzene 98%
Supplier: Apollo Scientific
|
Warning
|
OR4281-100G
OR4281-500G
OR4281-1G
APOSOR4281-100GEA
102
EUR
APOSOR4281-100G
APOSOR4281-500G
APOSOR4281-1G
1-Bromo-3-nitrobenzene 98%
1-Bromo-3-nitrobenzene
Formula:
C₆H₄BrNO₂ MW: 202,01 g/mol Boiling Pt: 256 °C Melting Pt: 51…54 °C Density: 1,704 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024298 CAS Number: 585-79-5 EINECS: 209-563-9 UN: 3459 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...