4-Bromoanisole 99%
Supplier: Apollo Scientific
|
Warning
|
OR4283-2.5KG
OR4283-100G
OR4283-500G
APOSOR4283-2.5KGEA
248
EUR
APOSOR4283-2.5KG
APOSOR4283-100G
APOSOR4283-500G
4-Bromoanisole 99%
4-Bromoanisole
Formula:
C₇H₇BrO MW: 187.04 g/mol Boiling Pt: 222…223 °C Melting Pt: 44478 °C Density: 1.494 g/cm³ (25 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000097 CAS Number: 104-92-7 EINECS: 203-252-1 Merck Index: 13,01412 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...