3'-Aminoacetophenone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
m-Aminoacetophenone, 1-(3-Aminophenyl)ethanone
OR4296-100G
OR4296-500G
APOSOR4296-100GEA
36.4
EUR
APOSOR4296-100G
APOSOR4296-500G
3'-Aminoacetophenone
3'-Aminoacetophenone
Formula:
C₈H₉NO MW: 135,17 g/mol Boiling Pt: 289…291 °C Melting Pt: 94…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007796 CAS Number: 99-03-6 EINECS: 202-722-3 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...